Cole-Parmer Technical Library
Ismatec™ Pump Selection Guide
Why Select Ismatec?    Selecting an Ismatec Pump   Selecting Ismatec Tubing
Pump System and Drive Selection by Flow Range
Why Select Ismatec?
  • Flow rates as low as 0.001 mL/min
  • High-accuracy numerical, digital, or programmable speed control
  • Tubing is the only wetted part
  • The quality of Swiss manufacturing
  • Most drives have a 2-year warranty
  • ISO 9000 Registered Supplier
  • CE compliant
Ismatec peristaltic pumps are available as complete systems or as separate drive and pump head components. A pump system includes a drive with a permanently attached pump head; a drive alone requires a separate pump head.
Remote control capabilities make Ismatec pumps ideal for lab automation. Most Ismatec drives offer remote on/off, speed, and directional control. Computer interfacing is possible with all digital drives.
A planetary drive design in which a central gear drives all rollers results in increased flow accuracy and tubing life. Rollers turn in unison to reduce stretching and slippage of tubing.
Selecting an Ismatec Pump
Flow Range depends on drive speed, number of rollers on the pump head, and the size of tubing. For a higher flow range, choose a pump head with fewer rollers. To reduce fluid pulsation, select a pump head with more rollers.
The number of channels refers to the number of pieces of tubing that can be used simultaneously. For example, a four-channel pump head can accommodate four pieces of tubing or four cartridges. Load the four channels with different types and or sizes of tubing then pump a variety of fluids or the same fluid at different flow rates.
Speed. Fixed-speed pumps and drives are simple and easy to use, yet they accept a wide variety of tubing sizes to deliver various flow rates.
Variable-speed pumps and drives accept analog input for remote on/off, speed, and directional control. They are available with numerical or digital display. Digital models provide an analog output to let you monitor flow rate and feature an RS-232 interface.
Selecting Ismatec Tubing
Look for the tubing information icons on each product page to determine at a glance what type of tubing to order for your pumps and pump heads. Flow rates will depend on both the rpm of your drive or pump and on the tubing diameter.
Pump System and Drive Selection by Flow Range
Model number Flow range
(mL/min)
Speed
(rpm)
Channels Rollers Tubing
type
Fixed-speed pumps (pump head included)
78012-00, -05 0.06 to 6.0 20 2 8 Three-stop
78011-10, -15 0.06 to 6.0 20 3 8 Three-stop
78012-30, -35 0.06 to 6.0 20 4 8 Three-stop
78015-30, -35 0.06 to 6.0 20 4 8 Three-stop
78011-20, -25 0.1 to 12.0 40 3 8 Three-stop
78012-40, -45 0.1 to 12.0 40 4 8 Three-stop
78012-20, -25 0.2 to 19.2 60 2 8 Three-stop
78011-30, -35 0.2 to 19.2 60 3 8 Three-stop
78015-00, -05 0.2 to 24.3 60 2 6 Three-stop
Variable-speed pumps with push-button potentiometer (pump head included)
78016-30 0.005 to 27.0 1 to 100 4 8 Three-stop
78022-40 0.005 to 150 3.5 to 350 8 6 Three-stop
78016-10 0.009 to 45.0 1.6 to 160 2 8 Three-stop
78006-00 0.01 to 44.4 2 to 120 8 to 40 6 Three-stop
78022-30 0.25 to 1200 3.5 to 350 1 3 Nominal
78022-20 1.6 to 5000 3.5 to 350 1 3 Nominal
78022-10 1.7 to 5400 3.5 to 350 1 2 Nominal
Variable-speed pumps with digital LCD display (pump head included)
78001-00, -10,
-20, -30, -40
78023-02, -12,
-22, -32, -42
0.001 to 8.86 0 to 11 4 to 24 8 Two-stop
78001-02, -12,
-22, -32, -42
78023-00, -10,
-20, -30, -40
0.004 to 35.4 0 to 45 4 to 24 8 Two-stop
78017-10 0.005 to 27.0 1 to 100 4 8 Three-stop
78017-00 0.009 to 45.0 1.6 to 160 2 8 Three-stop
78009-00, 78002-15 0.45 to 3400 1 to 240 1 3 Nominal
Variable-speed drives with push-button potentiometer
(pump head sold separately)
78007-00, -05 2 to 90
78005-10 2 to 200
78007-50, -55 4 to 240
78002-10 2 to 240
Variable-speed drives with digital LCD display (pump head sold separately)
78005-20 2 to 200
78002-00 2 to 240
Piston pumps (pump head included)
78021-00, -02 0.016 to 19
78021-04 to -14 0.064 to 77
78021-16 to -26 0.14 to 173
High-pressure piston pumps (pump head included)
78021-28 0.064 to 77
78021-30 0.14 to 173
78021-50 5 to 86
78021-52 10 to 172
Gear pump drive (pump heads sold separately)
78012-70 1 to 6540 60 to 600 1
Related Links: Printer-friendly page
Complete Selection of Ismatec Pumps
General Pump FAQs
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Terminology
Liquid Pumps Technical Equations and Information Tables
Pump Types and Definitions
Product Selection Guides  |  Product Maintenance  |  FAQs  |  Technical Library Map
Articles and White Papers  |  Case Studies  |  Conversion Factors and Tech Data